Methylcyclopentadiene dimer 95%
Leverantör: Thermo Scientific
Varning
156850010 156852500 156850025
ACRO156850010EA 2900 SEK
ACRO156850010 ACRO156852500 ACRO156850025
Methylcyclopentadiene dimer 95%
Methylcyclopentadiene dimer
Formel: C₁₂H₁₆
MW: 160,26 g/mol
Kokpunkt: 200 °C (760 mmHg)
Smältpunkt: –51 °C
Densitet: 0,94 g/cm³
Flampunkt: 26 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082297
CAS nummer: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
NMR (1H-NMR) ≥94 %