5-Hydroxy-1-tetralone ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
A17034.06 A17034.14
A17034.06EA 4675 SEK
A17034.06 A17034.14
5-Hydroxy-1-tetralone ≥99%
5-Hydroxy-1-tetralone
Purity: 99% CAS No.: 28315-93-7 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 209-211° Beilstein Reference: 2437410
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Smältpunkt: 209…211 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001693
CAS nummer: 28315-93-7
EINECS: 248-958-0