1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 99
Leverantör: Thermo Fisher Scientific
Synonyms: Beta-tetralone
L01053.06 L01053.14
L01053.06EA 1475 SEK
L01053.06 L01053.14
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 99
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone)
Purity: 99% CAS No.: 530-93-8 Molecular Formula: C10H10O Formula Weight: 146.19 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5600 Melting Point: 18° Boiling Point: 240° Beilstein Reference: 509386 MDL No.: MFCD00001727
Formel: C₁₀H₁₀O
MW: 146.19 g/mol
Kokpunkt: 240 °C
Densitet: 1.106
Flampunkt: >110 °C (230 °F)
MDL Number: MFCD00001727
CAS nummer: 530-93-8
EINECS: 208-498-3