2-Oxazolidinone 98%
Leverantör: Thermo Scientific
Synonyms: 2-Oxazolidone, Oxazolidin-2-one
129660250 129661000 129660050
ACRO129660250EA 805 SEK
ACRO129660250 ACRO129661000 ACRO129660050
2-Oxazolidinone 98%
2-Oxazolidinone
Formel: C₃H₅NO₂
MW: 87,08 g/mol
Kokpunkt: 220 °C (48 mmHg)
Smältpunkt: 86…89 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005268
CAS nummer: 497-25-6
EINECS: 207-840-9
Order Now
Specifikation testresultat
Appearance Off-white to yellow to light beige Powder or crystals
Infrared spectrum Conforms
GC ≥96.0 %