Hexamethyldisilathiane 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Bis(trimethylsilyl)sulphide
367370010 367370050 367370250
ACRO367370010EA 860 SEK
ACRO367370010 ACRO367370050 ACRO367370250
Hexamethyldisilathiane 95%
Hexamethyldisilathiane
Formel: C₆H₁₈SSi₂
MW: 178.45 g/mol
Kokpunkt: 163 °C (170 mmHg)
Densitet: 0.95 g/cm³
Flampunkt: 26 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014851
CAS nummer: 3385-94-2
EINECS: 222-201-4
UN: 1993
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %