Väteklorid 1 M i Dietyleter
Leverantör: Thermo Scientific
Fara
H27125.AE H27125.AP
H27125.AEEA 1600 SEK
H27125.AE H27125.AP
Väteklorid 1 M i Dietyleter
Väteklorid
CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Storage Temperature: 2°C to 8°C Flash Point: -40°(-40°F) Refractive Index: 1.3535 UN No.: 2924 MDL No.: MFCD00011324
Formel: HCl
MW: 36,46 g/mol
Densitet: 0,731
Flampunkt: –40 °C (–40 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3,I