Europium(III) sulphate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Varning
13210.06 13210.14
13210.06EA 3150 SEK
13210.06 13210.14
Europium(III) sulphate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Europium(III) sulphate octahydrate
Purity: 99.99% CAS No.: 10031-55-7 Molecular Formula: Eu2(SO4)3.8H2O Formula Weight: 736.23 (592.10anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00149704
Formel: Eu₂O₁₂S₃·8H₂O
MW: 736.23
Smältpunkt: 375 °C (-8H₂O)
Storage Temperature: Omgivande
MDL Number: MFCD00149703
CAS nummer: 10031-55-7
EINECS: 236-903-3
Merck Index: 14,3904