2-Chloro-5-propylpyrimidine

Leverantör: Thermo Scientific

RJC02409DA RJC02409EA RJC02409EE
MAYBRJC02409DAEA 0 SEK
MAYBRJC02409DA MAYBRJC02409EA MAYBRJC02409EE
2-Chloro-5-propylpyrimidine
2-Chloro-5-propylpyrimidine
Formel: C₇H₉ClN₂
MW: 156.61 g/mol
MDL Number: MFCD00799403
CAS nummer: 219555-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR