3-Octylthiophene ≥97%

Leverantör: Thermo Scientific
Fara

H55466.06 H55466.14
H55466.06EA 2725 SEK
H55466.06 H55466.14
3-Octylthiophene ≥97%
3-Octylthiophene

Purity: 97% CAS No.: 65016-62-8 Molecular Formula: C12H20S Formula Weight: 196.35 Flash Point: 113°(235°F) Physical Form: Liquid Density: 0.92 Refractive Index: 1.4920 Appearance: Colourless Boiling Point: 106-107°/3mm MDL No.: MFCD00085281

Formel: C₁₂H₂₀S
MW: 196,36 g/mol
Kokpunkt: 106…107 °C (3 mmHg)
Densitet: 0,92
Flampunkt: 113 °C (235 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00085281
CAS nummer: 65016-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR