1-Oktanol, anhydrous 99%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: n-Octanol
434581000 434580010
434581000.EA 845 SEK
434581000. 434580010.
1-Oktanol, anhydrous 99%, AcroSeal®
1-Oktanol
Formel: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Kokpunkt: 195 °C (760 mmHg)
Smältpunkt: –16 °C
Densitet: 0.824 g/cm³
Flampunkt: 81 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002988
CAS nummer: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 %
Residue after evaporation ≤0.004 %