3-(Trifluoromethyl)-1H-pyrazole ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-(Trifluoromethyl)-1H-pyrazole, 3-(Trifluoromethyl)pyrazole
H55068.03 H55068.06
H55068.03EA 1100 SEK
H55068.03 H55068.06
3-(Trifluoromethyl)-1H-pyrazole ≥97%
3-(Trifluoromethyl)-1H-pyrazole
Purity: 97% CAS No.: 20154-03-4 Molecular Formula: C4H3F3N2 Formula Weight: 136.08 Melting Point: 45-47° Boiling Point: 70°/2mm MDL No.: MFCD00115018
Formel: C₄H₃F₃N₂
MW: 136,08 g/mol
Kokpunkt: 70 °C (2 mmHg)
Smältpunkt: 45…47 °C
Storage Temperature: Omgivande
MDL Number: MFCD00115018
CAS nummer: 20154-03-4