Bortriklorid 1 M i hexane (mixed_isomers)
Leverantör: Thermo Fisher Scientific
Fara
H32833.AE H32833.AP
H32833.AEEA 1625 SEK
H32833.AE H32833.AP
Bortriklorid 1 M i hexane (mixed_isomers)
Bortriklorid
CAS No.: 10294-34-5 Molecular Formula: BCl3 Formula Weight: 117.17 Storage Temperature: 2°C to 8°C UN No.: 3286 MDL No.: MFCD00011313
Formel: BCl₃
MW: 117.17 g/mol
Kokpunkt: 12.4
Smältpunkt: -107
Densitet: 0.738
Storage Temperature: Kylskåp
MDL Number: MFCD00011313
CAS nummer: 10294-34-5
EINECS: 233-658-4
UN: 3286
ADR: 3 II
Merck Index: 12,01378