Tetrafluorophthalic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

T0986-25G T0986-5G
TCIAT0986-25GEA 750 SEK
TCIAT0986-25G TCIAT0986-5G
Tetrafluorophthalic acid ≥98.0%
Tetrafluorophthalic acid
Formel: C₈H₂F₄O₄
MW: 238,1 g/mol
Kokpunkt: 341 °C (1013 hPa)
Smältpunkt: 155…156 °C
MDL Number: MFCD00002407
CAS nummer: 652-03-9
EINECS: 211-483-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca8ee828c16645ab143ba0ba5c173d6f227a25c8&vid=ca8ee828c16645ab143ba0ba5c173d6f227a25c8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 149.0 to 156.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR