(3,5-Difluorophenyl)acetic acid

Leverantör: Thermo Scientific
Varning

Synonyms: 3,5-Difluorophenylacetic acid

SB01687EA
MAYBSB01687EAEA 0 SEK
MAYBSB01687EA
(3,5-Difluorophenyl)acetic acid
(3,5-Difluorophenyl)acetic acid
Formel: (C₆H₃F₂)CH₂COOH
MW: 172,13 g/mol
MDL Number: MFCD00010316
CAS nummer: 105184-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR