Oleinsyra ≥85.0%
Leverantör: TCI
Varning
Synonyms: (9Z)-9-Octadecenoic acid
O0011-25ML O0011-500ML
TCIAO0011-25MLEA 215 SEK
TCIAO0011-25ML TCIAO0011-500ML
Oleinsyra ≥85.0%
Oleinsyra
Formel: C₁₈H₃₄O₂
Kokpunkt: 285°C
Densitet: 0,8946 g/cm³ (20 °C)
Flampunkt: 187°C
Storage Temperature: Kylskåp
MDL Number: MFCD00064242
CAS nummer: 112-80-1
EINECS: 204-007-1
Merck Index: 14,06828
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc11a024921ec32ee758b9ad9c2b2e9993756b5&vid=efc11a024921ec32ee758b9ad9c2b2e9993756b5&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 85.0 %