Racks for micro tubes, floating , SP Bel-Art
Leverantör: Bel-Art Products, a Part of SP
F188756401 F188756411
390-6261EA 1300 SEK
390-6261 390-6266
Racks for micro tubes, floating , SP Bel-Art
Ställ Tube Racks
Molded of steam autoclavable polypropylene, these racks fit 400 ml or 1000 ml beakers.
  • For 1,5 ml micro tubes
  • Tubes remain completely immersed in a beaker while tops rest above the rack
  • Unload by lightly pressing down against the bench until the legs touch, releasing the tubes