Kontaktplattor, strålsteriliserade
Leverantör: VWR Chemicals
141114ZIEA 2625 SEK
141114ZI 141114PY 141114RY 141114TI 140884ZI 140884TI 140774ZI 140774TI 140070ZI 151203ZA 140884CY 141114XI
Kontaktplattor, strålsteriliserade
Media Kontaktplattor
Contact plates are ideal for monitoring microbiological contamination of surfaces and air (using an air sampler) inside cleanrooms, isolators, RABS (Restricted Access Barrier), the food industry and hospitals. Triple-wrapping ensures that the package itself does not contaminate the environment, as the first wrapper is removed just before entering a clean area. Plate packaging containing PCA is not irradiated.
  • Dubbel- eller trippelförpackning förhindrar kontaminering under transport och förvaring
  • Med strålnings-/dosimetricertifikat
  • Studier över VHP-resistens finns för alla trippelförpackade plattor
  • Förvaring vid rumstemperatur, 2-8 °C för TSA-penase
Förpackning: Contains 8×10 triple-wrapped packs with a total of 80 plates. 8 PET, transparent, reusable containers are included.