Nessler reagent, Supelco®
Leverantör: Merck
Fara
Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
M-Clarity™ quality level = MQ100
1.09028.0100 1.09028.0500
1.09028.0100EA 488 SEK
1.09028.0100 1.09028.0500
Nessler reagent, Supelco®
Nessler reagent
Determination of ammonium/nitrogen salts using mercury containing materials.
The classical colorimetric detection and determination of ammonium salts is carried out using Nessler's reagent- a yellow colour indicative of ammonium salts. A more recent development of the technique is to use a two-bottle system to prepare fresh reagent for each use.
MW: 786,4 g/mol
MDL Number: MFCD00049656
CAS nummer: 7783-33-7