Silikonolja, 20 cSt
Leverantör: VWR Chemicals
84543.360EA 3650 SEK
84543.360 84543.290
Silikonolja, 20 cSt
Silikonolja
Densitet: 0,95 g/cm³ (20 °C)
Flampunkt: 321 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00132673
CAS nummer: 63148-62-9
Order Now
Specifikation testresultat
Viscosity (25°C) 18 - 22 cSt
Volatile matter (150°C; 1 h) Max. 10.0 %