2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,2,6-Trimethyl-1,3-dioxine-4-one
296911000 296915000
ACRO296911000EA 1175 SEK
ACRO296911000 ACRO296915000
2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94%
2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Formel: C₇H₁₀O₃
MW: 142,15 g/mol
Kokpunkt: 65…67 °C (2,7mbar)
Smältpunkt: 4…13 °C
Densitet: 1,08 g/cm³
Flampunkt: 14 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040468
CAS nummer: 5394-63-8
EINECS: 226-403-3
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear yellow to dark brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4650 (20°C, 589 nm)
GC ≥92 %
Acetone ≤6 %