6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2,3,4-Tetrahydro-6,7-dimethoxyquinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-dione
L08411.09 L08411.18
L08411.09EA 1175 SEK
L08411.09 L08411.18
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥98%
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione
Purity: 98% CAS No.: 28888-44-0 Molecular Formula: C10H10N2O4 Formula Weight: 222.20 Melting Point: 300° Beilstein Reference: 204597
Formel: C₁₀H₁₀N₂O₄
MW: 222,2 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00023889
CAS nummer: 28888-44-0
EINECS: 249-288-1