2-Benzoxazolinone ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: benzo[d]oxazol-2(3H)-one
A19378.14 A19378.22
A19378.14EA 1125 SEK
A19378.14 A19378.22
2-Benzoxazolinone ≥98%
2-Benzoxazolinone
Purity: 98% CAS No.: 59-49-4 Molecular Formula: C7H5NO2 Formula Weight: 135.12 Flash Point: 160°(320°F) Melting Point: 139-140° Beilstein Reference: 119481 MDL No.: MFCD00005716
Formel: C₇H₅NO₂
MW: 135,12 g/mol
Smältpunkt: 139…140 °C
Flampunkt: 160 °C (320 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005716
CAS nummer: 59-49-4
EINECS: 200-430-0