Rhodanine-N-acetic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(4-Oxo-2-thioxothiazolidin-3-yl)acetic acid, Rhodanine-3-acetic acid
B22244.06 B22244.14 B22244.22
B22244.06EA 680 SEK
B22244.06 B22244.14 B22244.22
Rhodanine-N-acetic acid ≥98%
Rhodanine-N-acetic acid
Purity: 98% CAS No.: 5718-83-2 Molecular Formula: C5H5NO3S2 Formula Weight: 191.23 Melting Point: 145-148° Beilstein Reference: 149513
Formel: C₅H₅NO₃S₂
MW: 191,23 g/mol
Smältpunkt: 145…148 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005491
CAS nummer: 5718-83-2
EINECS: 227-220-1