Nickel aluminium alloy, pulver for production of Raney-nickel, för syntes, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: Nickel-aluminum alloy, Raney®-Nickel alloy, Aluminum-nickel alloy
M-Clarity™ quality level = MQ200
8.06749.0005 8.06749.0250 8.06749.1000
8.06749.0005EA 478 SEK
8.06749.0005 8.06749.0250 8.06749.1000
Nickel aluminium alloy, pulver for production of Raney-nickel, för syntes, Sigma-Aldrich®
Nickel aluminium alloy
for the production of Raney nickel
Melting Point : 1460 °C
Bulk Density: 2850 kg/m3
Density: 4.3 g/cm3
Formel: AlNi
MW: 85,675 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00167392
CAS nummer: 12635-29-9
EINECS: 234-439-6
UN: 1396
ADR: 4.3,III
Order Now
Specifikation testresultat
Assay of Ni 47.0 - 53.0 %
Assay of Al 47.0 - 53.0 %