Lithium tetrachlorocuprate 0.1 M i THF
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Dilithium tetrachlorocuprate
407830250 407831000 407835000
ACRO407830250EA 750 SEK
ACRO407830250 ACRO407831000 ACRO407835000
Lithium tetrachlorocuprate 0.1 M i THF
Lithium tetrachlorocuprate
Formel: Cl₄CuLi₂
MW: 219.24 g/mol
Densitet: 0.91 g/cm³
Flampunkt: –17 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011081
CAS nummer: 15489-27-7
UN: 1993
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear orange to red-brown Solution
Titration Argentometric 0.09 to 0.12 M
Titration with NaOH ≥95 %