Rhodanine 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one
152600250 152601000
ACRO152600250EA 910 SEK
ACRO152600250 ACRO152601000
Rhodanine 99%
Rhodanine
Formel: C₃H₃NOS₂
MW: 133,19 g/mol
Smältpunkt: 167…170 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005488
CAS nummer: 141-84-4
EINECS: 205-505-1
Order Now
Specifikation testresultat
Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 167 to 170 °C
Titration with NaOH ≥98.5 %