Potassium-2,5-dihydroxybenzenesulphonate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate
L08406.22 L08406.36
L08406.22EA 825 SEK
L08406.22 L08406.36
Potassium-2,5-dihydroxybenzenesulphonate ≥98%
Potassium-2,5-dihydroxybenzenesulphonate
Purity: 98+% CAS No.: 21799-87-1 Molecular Formula: C6H5KO5S Formula Weight: 228.27 Melting Point: ca 251° dec. Beilstein Reference: 5187027
Formel: C₆H₅KO₅S
MW: 228,27 g/mol
Smältpunkt: ∼ 251 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007475
CAS nummer: 21799-87-1
EINECS: 244-584-7