Trimellitic anhydride ≥97%
Leverantör: Thermo Fisher Scientific
Fara
B23003.30 B23003.A1
B23003.30EA 690 SEK
B23003.30 B23003.A1
Trimellitic anhydride ≥97%
Trimellitic anhydride
Purity: 97% CAS No.: 552-30-7 Molecular Formula: C9H4O5 Formula Weight: 192.13 Flash Point: 227°(440°F) Melting Point: 165-169° Boiling Point: 240-245°/14mm Beilstein Reference: 9394 Merck Reference: 14,9703 MDL No.: MFCD00005925
Formel: C₉H₄O₅
Kokpunkt: 240…245 °C (14 mmHg)
Smältpunkt: 165…169 °C
Densitet: 1,540
Flampunkt: 227 °C (440 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005925
CAS nummer: 552-30-7
EINECS: 209-008-0
Merck Index: 14,9703