Bis(methylthio)methane ≥99%
Leverantör: Thermo Scientific
Varning
A19706.06 A19706.14
A19706.06EA 710 SEK
A19706.06 A19706.14
Bis(methylthio)methane ≥99%
Bis(methylthio)methane
Purity: 99% CAS No.: 1618-26-4 Molecular Formula: CH3SCH2SCH3 Formula Weight: 108.23 Flash Point: 43°(109°F) Refractive Index: 1.5340 Odour: Unpleasant Boiling Point: 146-147° Beilstein Reference: 1731143 Merck Reference: 14,1256 UN No.: 1993 MDL No.: MFCD00008564
Formel: C₃H₈S₂
MW: 108,23 g/mol
Kokpunkt: 146…147 °C
Densitet: 1,059
Flampunkt: 43 °C (109 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008564
CAS nummer: 1618-26-4
EINECS: 216-577-9
ADR: 3,III
Merck Index: 14,1256