Sodium mercaptoacetate (sodium thioglycolate) ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt
L14356.14 L14356.22 L14356.36
L14356.14EA 650 SEK
L14356.14 L14356.22 L14356.36
Sodium mercaptoacetate (sodium thioglycolate) ≥97%
Sodium mercaptoacetate (sodium thioglycolate)
Purity: 97% CAS No.: 367-51-1 Molecular Formula: HSCH2CO2Na Formula Weight: 114.10 Odour: Stench pH: ~6 Melting Point: ca 305° Beilstein Reference: 4569109 Merck Reference: 14,8692 UN No.: 3335 MDL No.: MFCD00043386
Formel: HSCH₂COONa
MW: 114,1 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00043386
CAS nummer: 367-51-1
EINECS: 206-696-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,8692