Calcium thioglycolate trihydrate, crystallised
Leverantör: Merck
Varning
Synonyms: Calcium mercaptoacetate trihydrate, Calcium thioglycolate trihydrate, Calcium bis(mercaptoacetate) trihydrate, Mercaptoacetic acid calcium salt trihydrate
1.02125.1000 1.02125.9075
1.02125.1000EA 0 SEK
1.02125.1000 1.02125.9075
Calcium thioglycolate trihydrate, crystallised
Calcium thioglycolate trihydrate
Formel: C₄H₆CaO₄S₂·3H₂O
MW: 276.35 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00150707
CAS nummer: 65208-41-5
EINECS: 249-881-5
UN: 3263
ADR: 8 III