Lithium-tert-butanolate 2.2 M i THF, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
379311000 379318000
379311000.EA 1800 SEK
379311000. 379318000.
Lithium-tert-butanolate 2.2 M i THF, AcroSeal®
Lithium-tert-butanolate
Formel: C₄H₉LiO
MW: 80.06 g/mol
Flampunkt: –22 °C
Storage Temperature: Omgivande
MDL Number: MFCD00050479
CAS nummer: 1907-33-1
EINECS: 217-611-5
UN: 2924
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear yellow to brown Solution
Strength 19 to 21 %
Impurity ≤0.2 % LiOH + Li2CO3 (as LiOH)