2-Chloro-4,6-dimethylpyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

C2199-25G C2199-5G
TCIAC2199-25GEA 1875 SEK
TCIAC2199-25G TCIAC2199-5G
2-Chloro-4,6-dimethylpyrimidine ≥98.0%
2-Chloro-4,6-dimethylpyrimidine
Formel: C₆H₇ClN₂
MW: 142,59 g/mol
Kokpunkt: 212°C
Storage Temperature: Kylskåp
MDL Number: MFCD00023199
CAS nummer: 4472-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cfacd12633674fd6cbd749dc1369c631af0dac0&vid=3cfacd12633674fd6cbd749dc1369c631af0dac0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR