2-Chloro-4,6-dimethoxypyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

C1433-10G C1433-25G
TCIAC1433-10GEA 215 SEK
TCIAC1433-10G TCIAC1433-25G
2-Chloro-4,6-dimethoxypyrimidine ≥98.0%
2-Chloro-4,6-dimethoxypyrimidine
Formel: C₆H₇ClN₂O₂
MW: 174,58 g/mol
MDL Number: MFCD00274530
CAS nummer: 13223-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bc6df7598338da76af4ac73fbb82b84ae2fbb02&vid=7bc6df7598338da76af4ac73fbb82b84ae2fbb02&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR