Methyl phenyl sulphoxide 98+%
Leverantör: Thermo Scientific
Synonyms: (Methylsulphinyl)benzene
352120050 352120250
ACRO352120050EA 925 SEK
ACRO352120050 ACRO352120250
Methyl phenyl sulphoxide 98+%
Methyl phenyl sulphoxide
Formel: C₇H₈OS
MW: 140,21 g/mol
Kokpunkt: 92…94 °C (,3 mmHg)
Smältpunkt: 34…36 °C
Flampunkt: 86 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002088
CAS nummer: 1193-82-4
EINECS: 214-781-2
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid or fused mass
Infrared spectrum Conforms
Refractive index 1.5755 to 1.5795 (20°C, 589 nm)
GC ≥98.0 %