5-Amino-1-methyl-1H-pyrazole

Leverantör: Thermo Scientific

Synonyms: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

GK03066DA GK03066EA
MAYBGK03066DAEA 0 SEK
MAYBGK03066DA MAYBGK03066EA
5-Amino-1-methyl-1H-pyrazole
5-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
MW: 97,12 g/mol
MDL Number: MFCD00068156
CAS nummer: 1192-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR