3-Amino-1H-pyrazole 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole
171030025 171030100
ACRO171030025EA 1275 SEK
ACRO171030025 ACRO171030100
3-Amino-1H-pyrazole 98%
3-Amino-1H-pyrazole
Formel: C₃H₅N₃
MW: 83,09 g/mol
Kokpunkt: 218 °C (122 mmHg)
Smältpunkt: 33…39 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005236
CAS nummer: 1820-80-0
EINECS: 217-338-1
UN: 3263
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to light brown Liquid or fused solid
Infrared spectrum Conforms
GC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %