Coumarin-3-carboxylic acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Oxo-2H-chromene-3-carboxylic acid
346760250 346761000
ACRO346760250EA 895 SEK
ACRO346760250 ACRO346761000
Coumarin-3-carboxylic acid 98%
Coumarin-3-carboxylic acid
Formel: C₁₀H₆O₄
MW: 190,16 g/mol
Smältpunkt: 190…193 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006852
CAS nummer: 531-81-7
EINECS: 208-518-0
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥97.5 %