2,2,6,6-Tetramethylpiperidinooxy (TEMPO) 98%
Leverantör: Thermo Scientific
Fara
Synonyms: TEMPO
200380050 200380250
ACRO200380050EA 765 SEK
ACRO200380050 ACRO200380250
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) 98%
2,2,6,6-Tetramethylpiperidinooxy (TEMPO)
Formel: C₉H₁₈NO
MW: 156,25 g/mol
Smältpunkt: 36…40 °C
Flampunkt: 67 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009599
CAS nummer: 2564-83-2
EINECS: 219-888-8
UN: 3263
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Orange to red Crystalline powder or crystals or solid and/or chunks
Infrared spectrum Conforms
GC ≥97.5 %