Diethyl phenyl orthoformate ≥97%
Leverantör: Thermo Scientific
Varning
B21498.09 B21498.18
B21498.09EA 1150 SEK
B21498.09 B21498.18
Diethyl phenyl orthoformate ≥97%
Diethyl phenyl orthoformate
Purity: 97% CAS No.: 14444-77-0 Molecular Formula: C11H16O3 Formula Weight: 196.25 Flash Point: 65°(149°F) Refractive Index: 1.4810 Melting Point: -26° Boiling Point: 103-104°/10mm Beilstein Reference: 1872634 MDL No.: MFCD00009228
Formel: C₁₁H₁₆O₃
MW: 196,25 g/mol
Kokpunkt: 103…104 °C (10 mmHg)
Smältpunkt: –26 °C
Densitet: 1,015
Flampunkt: 65 °C (149 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009228
CAS nummer: 14444-77-0
EINECS: 238-421-9