Methyl isobutyrylacetate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
L16216.14 L16216.22
L16216.14EA 1125 SEK
L16216.14 L16216.22
Methyl isobutyrylacetate ≥98%
Methyl isobutyrylacetate
Purity: 98% CAS No.: 42558-54-3 Molecular Formula: C7H12O3 Formula Weight: 144.17 Flash Point: 79°(174°F) Refractive Index: 1.4265 Melting Point: -75° Boiling Point: 192° Beilstein Reference: 1857823 MDL No.: MFCD00040499
Formel: C₇H₁₂O₃
Kokpunkt: 192 °C
Smältpunkt: –75 °C
Densitet: 1,010
Flampunkt: 79 °C (174 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00040499
CAS nummer: 42558-54-3
EINECS: 418-900-0