Methoxyhydroquinone ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Methoxyhydroquinone
A12847.09 A12847.18 A12847.30
A12847.09EA 930 SEK
A12847.09 A12847.18 A12847.30
Methoxyhydroquinone ≥97%
Methoxyhydroquinone
Purity: 97% CAS No.: 824-46-4 Molecular Formula: C7H8O3 Formula Weight: 140.14 Melting Point: 87-91° Beilstein Reference: 2045285
Formel: C₇H₈O₃
MW: 140,14 g/mol
Smältpunkt: 87…91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013971
CAS nummer: 824-46-4
EINECS: 212-530-1