4-Piperidinemethanol ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Piperidin-4-ylmethanol
L17964.06 L17964.14
L17964.06EA 2200 SEK
L17964.06 L17964.14
4-Piperidinemethanol ≥97%
4-Piperidinemethanol
Purity: 97% CAS No.: 6457-49-4 Molecular Formula: C6H13NO Formula Weight: 115.18 Flash Point: 110°(230°F) Odour: Amine-like Boiling Point: 118-120°/10mm Beilstein Reference: 103071 UN No.: 3259 MDL No.: MFCD00174228
Formel: C₆H₁₃NO
MW: 115,18 g/mol
Kokpunkt: 118…120 °C (10 mmHg)
Smältpunkt: 57…61 °C
Densitet: 0,997
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00174228
CAS nummer: 6457-49-4
ADR: 8,III