3-Piperidinemethanol 96%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-(Hydroxymethyl)piperidine
L04264.06 L04264.14
L04264.06EA 725 SEK
L04264.06 L04264.14
3-Piperidinemethanol 96%
3-Piperidinemethanol
Purity: 96% CAS No.: 4606-65-9 Molecular Formula: C6H13NO Formula Weight: 115.18 Flash Point: 110°(230°F) Refractive Index: 1.4940 Melting Point: 58-62° Boiling Point: 236° Beilstein Reference: 103058 UN No.: 3259 MDL No.: MFCD00005997
Formel: C₆H₁₃NO
MW: 115,18 g/mol
Kokpunkt: 236 °C
Smältpunkt: 58…62 °C
Densitet: 1,025
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005997
CAS nummer: 4606-65-9
EINECS: 225-013-0
ADR: 8,III