Bly standardlösning, 1000 mg/L Pb in 5% salpetersyra (av Pb) Specpure® standard för ICP
Leverantör: Thermo Fisher Scientific
13853.AD 13853.AE 13853.AP 14412.AD 14412.AE 14412.AP
13853.ADEA 1325 SEK
13853.AD 13853.AE 13853.AP 14412.AD 14412.AE 14412.AP
Bly standardlösning, 1000 mg/L Pb in 5% salpetersyra (av Pb) Specpure® standard för ICP
Standarder Cationic Standards Lead Standards
Pb in 5% HNO₃
UN No.: 3264
MDL No.: MFCD00134050
Certifikat: Traceable to NIST