Ethyl mercaptoacetate 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ethyl 2-mercaptoacetate, Ethyl thioglycolate
118671000 118675000 118670025
ACRO118671000EA 1175 SEK
ACRO118671000 ACRO118675000 ACRO118670025
Ethyl mercaptoacetate 99%
Ethyl mercaptoacetate
Formel: C₄H₈O₂S
Kokpunkt: 54 °C (12 mmHg)
Smältpunkt: –80 °C
Densitet: 1,09 g/cm³
Flampunkt: 47 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004874
CAS nummer: 623-51-8
EINECS: 210-800-3
UN: 1228
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4590 (20°C, 589 nm)
GC ≥98.5 %