Ethyl oxalyl chloride 98%
Leverantör: Thermo Fisher Scientific
Fara
118780250 118781000 118785000
ACRO118780250EA 605 SEK
ACRO118780250 ACRO118781000 ACRO118785000
Ethyl oxalyl chloride 98%
Ethyl oxalyl chloride
Formel: C₄H₅ClO₃
Kokpunkt: 135 °C (760 mmHg)
Densitet: 1,22 g/cm³
Flampunkt: 41 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000706
CAS nummer: 4755-77-5
EINECS: 225-285-0
UN: 2920
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤200 APHA