Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Dibenzo-18-crown-6
A13133.06 A13133.14 A13133.22
A13133.06EA 780 SEK
A13133.06 A13133.14 A13133.22
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥98%
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I)
Purity: 98+% CAS No.: 14187-32-7 Molecular Formula: C20H24O6 Formula Weight: 360.41 Melting Point: 160-164° Boiling Point: 380-384°/679mm Merck Reference: 14,2602 MDL No.: MFCD00005098
Formel: C₂₀H₂₄O₆
MW: 360,41 g/mol
Kokpunkt: 380…384 °C (679 mmHg)
Smältpunkt: 160…164 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005098
CAS nummer: 14187-32-7
EINECS: 238-041-3
Merck Index: 14,2602