Bis(2-ethylhexyl)phosphate 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid
402470010 402470050
ACRO402470010EA 4125 SEK
ACRO402470010 ACRO402470050
Bis(2-ethylhexyl)phosphate 95%
Bis(2-ethylhexyl)phosphate
Formel: C₁₆H₃₅O₄P
MW: 322.43 g/mol
Kokpunkt: 48 °C (12 mmHg)
Smältpunkt: –60 °C
Densitet: 0.97 g/cm³
Flampunkt: 137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009492
CAS nummer: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
Titration with NaOH ≥94 %