p-Phenylenediacetonitrile ≥97%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
2,2'-(1,4-Phenylene)diacetonitrile, 1,4-Phenylenediacetonitrile
L13464.06
L13464.14
L13464.06EA
875
SEK
L13464.06
L13464.14
p-Phenylenediacetonitrile ≥97%
p-Phenylenediacetonitrile
Purity: 97%
CAS No.: 622-75-3
Molecular Formula: C10H8N2
Formula Weight: 156.19
Melting Point: 94-98°
Beilstein Reference: 1866157
UN No.: 3439
MDL No.: MFCD00001923
Formel:
C₁₀H₈N₂ MW: 156,19 g/mol Smältpunkt: 94…98 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00001923 CAS nummer: 622-75-3 EINECS: 210-751-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...